
Calcium chloride, anhydrous ≥96.0% (by argentometric titration), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Merck Index:14,01659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2.174 g/cm³ (20 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
Specification Test Results
- Assay (CaCl₂) (by Ag titrn):≥96.0 %
- Titrable Base (meq/g):≤0.006
- Mesh - On U.S. No. 20 Sieve:≤10 %
- Mesh - Thru U.S. No. 200 Sieve:≤30 %
Specification Test Results
Related Information
Frequently Bought Together