Order Entry
Switzerland
ContactUsLinkComponent
Aluminium potassium sulphate dodecahydrate 98.0-102.0%, granular, AR® ACS, Macron Fine Chemicals™
Aluminium potassium sulphate dodecahydrate 98.0-102.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 3216-20
CAS Number:  
undefined
Aluminium potassium sulphate dodecahydrate 98.0-102.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 3216-20
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 102.0 %
  • ACS - Assay (AlK(SO₄)₂·12H₂O):
    98.0 - 102.0 %
  • ACS - Ammonium (NH₄):
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤0.0005 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - pH of 5% Solution at 25°C:
    2.0 - 4.0
  • ACS - Sodium (Na):
    ≤0.02 %
  • Solubility:
    Passes Test

Specification Test Results

About this item

 :
 :474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.72 g/cm³ (20 °C)
EINECS:233-141-3
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143