Order Entry
Switzerland
ContactUsLinkComponent
 
Acetone, Finyte® for microelectronic, J.T.Baker®
Catalog # 5018-20
Supplier:  Avantor
CAS Number:  
undefined
Acetone, Finyte® for microelectronic, J.T.Baker®
Catalog # 5018-20
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    330 lb
  • Seq:
    7242

Specifications

Specification Test Results

  • For Microelectronic Use:
    = 99.5 %
  • Assay (by GC):
    ≥ 99.5 %
  • Color (APHA):
    ≤ 10
  • Acidity (µeq/g):
    ≤ 0.3
  • Aldehydes (as HCHO):
    ≤ 20000 ppb
  • Alkalinity (µeq/g):
    ≤ 0.5
  • Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):
    ≤ 0.05 %
  • Methanol (CH₃OH)(by GC):
    ≤ 0.05 %
  • Sulfate (SO₄), For Information Only:
    = 3000.0 ppb
  • Residue after Evaporation:
    ≤ 3000.0 ppb
  • Water (H₂O):
    ≤ 0.50 %
  • Solubility in H₂O:
    Passes Test
  • Chloride (Cl):
    ≤ 200.0 ppb
  • Phosphate (PO₄):
    ≤ 50.0 ppb
  • Trace Impurities - Aluminum (Al):
    ≤ 10.0 ppb
  • Trace Impurities - Antimony (Sb):
    ≤ 5.0 ppb
  • Trace Impurities - Arsenic (As):
    ≤ 5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤ 10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤ 10.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 10.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 10.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 10.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤ 10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤ 10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 10.0 ppb
  • Trace Impurities - Silicon:
    ≤ 10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 10.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤ 10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤ 10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤ 10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 10.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤ 10.0 ppb
  • Particle Count at point of fill - 0.5 µm and greater:
    ≤ 150 par/ml
  • Particle Count at point of fill - 1.0 µm and greater:
    ≤ 25 par/ml

Specification Test Results

About this item

 :
 :58.08 g/mol
UN:1090
ADR:3,II
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765