

Acetone ≥99.5%, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Catalog # 9009-07
Supplier: Avantor
CAS Number:
Specifications
- Pk:5 gal US
- Seq:8160
Specifications
Specification Test Results
- For Microelectronic Use:= 99.5 %
- Assay ((CH₃)₂CO) (by GC, corrected for water):≥ 99.5 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 0.001 %
- Solubility in H₂O:Passes Test
- Titrable Acid (meq/g):≤ 0.0003
- Titrable Base (meq/g):≤ 0.0006
- Aldehydes (as HCHO):≤ 0.002 %
- Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):≤ 0.05 %
- Methanol (CH₃OH)(by GC):≤ 0.05 %
- Heavy Metals (as Pb):≤ 0.20 ppm
- Substances Reducing Permanganate:Passes Test
- Water (H₂O):≤ 0.5 %
- Chloride (Cl):≤ 0.2 ppm
- Copper (Cu):≤ 0.1 ppm
- Iron (Fe):≤ 0.050 ppm
- Nickel (Ni):≤ 0.1 ppm
- Manganese (Mn):≤ 0.100 ppm
Specification Test Results
About this item
:
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765