Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Acetone ≥99.5% (by GC, corrected for water content), AR® ACS for histology, Macron Fine Chemicals™
  2440-24
 :  
undefined
 
 
 
Acetone ≥99.5% (by GC, corrected for water content), AR® ACS for histology, Macron Fine Chemicals™
  2440-24
 :  

 

  • Pack type:
    Steel drum
  • Pk:
    350 lb
  • Seq:
    7251

 

 

  • Meets ACS Reagent Chemical Requirements:
    ≥99.5%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.002%
  • ACS - Assay (CH₃COCH₃) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Aldehydes (as HCHO):
    ≤0.002 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):
    ≤0.05 %
  • ACS - Methanol (CH₃OH)(by GC):
    ≤0.05 %
  • ACS - Residue after Evaporation:
    ≤0.001 %
  • ACS - Solubility in H₂O:
    Passes Test
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • ACS - Titrable Acid (meq/g):
    ≤0.0003
  • ACS - Titrable Base (meq/g):
    ≤0.0006
  • ACS - Water (H₂O):
    ≤0.5 %

 

 

 :
 :58.08 g/mol
 :13,00067
 :56.2 °C (1013 hPa)
 :–95.4 °C
 :0.792 g/cm³ (20 °C)
 :≤20 °C (closed cup)
 :Ambient
 :200-662-2
 :1090
 :3,II
 :01-2119471330-49
 :GHS02
 :GHS07
 :GHS08
 :67-64-1
 :CH₃COCH₃
 :MFCD00008765