Organic reference standards, fexofenadines, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:185066-37-9
- Formula:C₃₁H₃₉NO₂
- Description:Fexofenadine impurity C, (1RS)-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]-1-[4-(1-methylethyl)phenyl]butan-1-ol
Specifications
Frequently Bought Together