Anti-S100B Mouse Monoclonal Antibody (AF488) [clone: S100B/1012]
ABNOMAB15239
: Abnova
- Antibody type:Primary
- Antigen symbol:S100B
- Clonality:Monoclonal
- Clone:S100B/1012
- Conjugation:Alexa Fluor® 488
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:6285
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Storage temperature:Store at 4 °C.
- Immunogen:Recombinant protein corresponding to full length human S100B.
- Purification:Protein A/G purification
- Size:500 µL
- Pk:500 µl
Mouse monoclonal antibody raised against full length recombinant human S100B.
Type: Primary
Antigen: S100B
Clonality: Monoclonal
Clone: S100B/1012
Conjugation: AF488
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human