Order Entry
Switzerland
ContactUsLinkComponent
Taq DNA polymerase, 1000 U
Taq DNA polymerase, 1000 U
Catalog # GENSE00008
Supplier:  Genscript
CAS Number:  
Taq DNA polymerase, 1000 U
Catalog # GENSE00008
Supplier:  Genscript
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Taq DNA polymerase, 1000 units

Specifications

About this item

Taq DNA polymerase is a thermostable DNA polymerase isolated from an E. coli strain that carries the Taq DNA polymerase gene.

  • Used for PCR reactions
  • Terminal transferase activity
  • High purity

One unit is the amount of enzyme that can incorporate 10 nmol of dNTP into acid-insoluble material in 30 minutes at 74 °C.