Order Entry
Switzerland
ContactUsLinkComponent
 
EDTA solution, 0,5 M, molecular biology grade, Calbiochem®, Millipore®
Katalognr. 324506-100
CAS-Nummer:  
EDTA solution, 0,5 M, molecular biology grade, Calbiochem®, Millipore®
Katalognr. 324506-100
CAS-Nummer:  

Technische Daten

  • Pk:
    100 mL
  • Description:
    EDTA solution

Technische Daten

Über diesen Artikel

Sterile-filtered solution of 0,5 M EDTA in H₂O treated with diethyl pyrocarbonate (DEPC). Used in molecular biology applications.

  • pH: 8,0
  • Diethyl pyrocarbonate (DEPC) treated

Synonyms: Ethylenediaminetetraacetic acid trisodium salt.