Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Wall mounted thermometer

Wall mounted thermometer

Analog wall thermometer, plastic, 400×68 mm, −30…+50 °C
Catalog # MOIN3005YO
Supplier:   MOINEAU
Wall mounted thermometer
Catalog # MOIN3005YO
Supplier:  MOINEAU
Wall mounted thermometer
Wall mounted thermometer

Analog wall thermometer, plastic, 400×68 mm, −30…+50 °C

Price
SFr. 13.70
Log in for availability
1 * 1 ST
Product Details & Documents

Large indoor/outdoor thermometer with alcohol filling.

  • Made from white ABS
  • With hole for wall mounting
Specifications
  • Description:
    Analog wall thermometer, plastic, 400×68 mm
  • Range:
    −30…+50 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.