Ethylene oxide sterilisation indicators
The discs are available in a variety of blank or printed versions.
Sterilisation occurs on exposure to ethylene oxide (ETO).
The 489-0097 indicator changes colour from purple to green when exposed to ETO.
The 489-0098 indicator carries the text: 'Green is ETO exposed'.
: Roll of 5000 labels.
Frequently Bought Together