Fit test for respirators, FT10
: 3M
Test kits and accessories designed to help your organisation meet OSHA fit testing requirements for respirators. The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapour respirator with a particulate prefilter. FT-10 uses a test solution of sodium saccharin that produces a sweet tasting aerosol.
- Qualitative fit test kits are suitable for testing wearers of disposable respirators and half face masks fitted with particle filters
- Kits rely on a ‘taste test’ to assess the level of fit of the respirator to the wearer’s face
For 3M respirators: 8812, 9322, 8835, 4000 series 6000 series, 7500 series.
: Respirator should be fit tested, as required under the Control of Substances Hazardous to Health Regulations (COSHH). The kit meets the performance criteria for fit testing respirators under the current OSHA Standard for Respirator Protection: 29 CFR 1910.134, Appendix A.
: Kit includes: hood, collar, 2 nebulisers, sweet tasting sensitivity solution, sweet tasting fit test solution and laminated user instructions.
Frequently Bought Together