Order Entry
Switzerland
Orders LinkContactUsLinkComponent
<i>Trichoderma</i> endo-1,3-beta-D-glucanase (from <i>Trichoderma</i> sp.)

Trichoderma endo-1,3-beta-D-glucanase (from Trichoderma sp.)

100 U, 100
Catalog # MEGAE-LAMSE
Supplier:   MEGAZYME
<i>Trichoderma</i> endo-1,3-beta-D-glucanase (from <i>Trichoderma</i> sp.)
Catalog # MEGAE-LAMSE
Supplier:  MEGAZYME
CAS Number:  
Trichoderma endo-1,3-beta-D-glucanase (from Trichoderma sp.)

100 U, 100

CAS Number:  
Price
SFr. 363.00
Log in for availability
1 * 100 ST
Product Details & Documents

endo-1,3-β-D-Glucanase is isolated from Trichoderma species. These are used in carbohydrate and biofuels research, biochemical enzyme assays and in vitro diagnostic analysis.

  • CAS.No: 9025-37-0
  • Form: In 3,2 M ammonium sulphate
  • Stability: >4 years at 4 °C
  • Specific activity: ~15 U/mg (40 °C, pH 4,5 on CM-Curdian)
  • Specificity: Hydrolysis of (1,3)-β-D-glucosidic linkages in (1,3)-β-D-glucans

One unit of endo-1,3-β-D-Glucanase activity is defined as the amount of enzyme required to release one μmole of glucose-reducing-sugar equivalents per minute in the presence of CM-Curdlan (5 mg/ml) in sodium acetate buffer (200 mM) at pH 4,5 and 40 °C.

Specifications

Specifications

  • Pk:
    100
  • Pk Info:
    100 U
  • Cat. No.:
    MEGAE-LAMSE
  • Storage conditions:
    4 °C
  • Species:
    Trichoderma
  • Source:
    Trichoderma sp.
  • Enzyme name:
    endo-1,3-β-D-Glucanase

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.