ContactUsLinkComponent
Multi-gas detector, Accuro®
Multi-gas detector, Accuro®
  DRAE8318392
 :  Dräger
Multi-gas detector, Accuro®
  DRAE8318392
 :  Dräger
  • Model:
    Accuro®
  • Description:
    Multi-gas detector
  • Pk:
    1
Specifically designed for fast measurements with one hand, this gas detector pump allows to use tested tubes under extreme conditions.
 : Includes hand bellows pump and spares kit in standard leather carrying case.