ContactUsLinkComponent
Adhesive seal, AMPLIseal™
Adhesive seal, AMPLIseal™
Catalog # 736-0354
Supplier:  Greiner Bio-One
undefined
Adhesive seal, AMPLIseal™
Catalog # 736-0354
Supplier:  Greiner Bio-One
Specifications
  • Optical property:
    Transparent
  • Description:
    AMPLIseal™
  • Pk:
    100
Specifications
About this item
AMPLIseal™ is a self-adhesive sealing film notable for its low autofluorescence, in particular in the wavelength range critical for Real Time PCR. It consists of a thick PP film coated with a transparent acrylate adhesive. The adhesive layer is protected by a peelable polyester film. The strongly adhering acrylate adhesive provides reliable sealing, thereby minimising evaporation but without influencing the PCR reaction or the fluorescence measurement.
  • Highly transparent with minimal autofluorescence
  • Ideal for Real Time PCR