Order Entry
Switzerland
ContactUsLinkComponent
 
EDTA disodium salt solution, 0,2% in phosphate buffered saline, MP Biomedicals
  ICNA092820549
 :  
EDTA disodium salt solution, 0,2% in phosphate buffered saline, MP Biomedicals
  ICNA092820549
 :  

 

  • Pk:
    100 mL
  • Description:
    EDTA Disodium salt 2%

 

 

EDTA disodium salt, (2%) solution, in phosphate buffered saline without calcium and magnesium. Used as a gentle non-enzymatic cell dissociation reagent.

  • Clear and colourless
  • pH: 4,0 to 6,0
  • Osmolality: 240 to 450 mM/kg