Avantor® Apollo Silica, HPLC Columns
: Avantor
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations.
- Easy scale-up from analytical to prep
- Suitable for pH 2 - 8
- USP L3
These columns are ideal for routine analysis and for educational laboratories.
- Catalog No:
- 96419/N
- HICH36507
- HICH36513
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)