Order Entry
Switzerland
ContactUsLinkComponent
Dehydrated culture media, SOC medium, MP Biomedicals
Dehydrated culture media, SOC medium, MP Biomedicals
Catalog # ICNA113031012
Supplier:  MP Biomedicals
CAS Number:  
undefined
Dehydrated culture media, SOC medium, MP Biomedicals
Catalog # ICNA113031012
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    227 g
  • Description:
    SOC medium, powder
  • Storage conditions:
    Store at room temperature
  • Sterility:
    0,2 μm filtered
  • Application:
    Bacteria Cell Culture; Transformation
  • pH:
    6,5 ±0,2
  • Format:
    Powder

Specifications

About this item

SOC medium is a rich media used primarily in the recovery step of Escherichia coli competent cell transformations.

  • Maximises the transformation efficiency of competent cells

SOC powder is a special medium for incubating competent cells immediately after transformation to allow expression of transferred resistance genes before exposing cells to selective conditions.

Ordering information: Small quantities of biological buffers have been added for proper pH adjustment.