ContactUsLinkComponent
Dehydrated culture media, LB-medium, molecular biology grade, MP Biomedicals
Dehydrated culture media, LB-medium, molecular biology grade, MP Biomedicals
Nome del fornitore:  MP Biomedicals
Certificati
undefined
Dehydrated culture media, LB-medium, molecular biology grade, MP Biomedicals
Nome del fornitore:  MP Biomedicals
Certificati
Specifiche
  • Storage temperature:
    Store sterile LB Agar Medium at ambient temperature (15 - 30 °C). Powder medium is hygroscopic, therefore minimize exposure to air to prolong shelf life of media
Specifiche
Informazioni correlate
Frequently Bought Together