Accessories for autoclaves Vacuum Pro CertoClav series
Frequently Bought Together
PLUG, BOTTLE 5/8 (G) 1 * 1 items
: Beckman Coulter
Precision laboratory thermometers
: Ludwig Schneider GmbH & CO. KG
Accessories for tray dryers, FreeZone®
: LABCONCO
Infrared sterilizer Model with 14 mm diameter inside 1 * 1 ST
: MARKET SOURCE PART PROCESS
808 nm, 120 mW, APC, TTL 5 kHz, focusable, 5 VDC, 22 x 65 mm 1 * 1 ST
: MARKET SOURCE PART PROCESS
Mesh Stainless Steel Locker
: TEKNOMEK
Stainless Steel Wire Modular Shelving Unit
: TEKNOMEK
BOTTLES, PP 4 X 6-1/2 (BOX-6) 1 * 6 Bottle
: Beckman Coulter
BAKTOLIN FOAM CLEAN SAFE 1L 1 * 6 items
: BODE CHEMIE