Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Corning® Trypsin

Corning® Trypsin

Supplier:   Corning
Corning® Trypsin
Supplier:  Corning
Certificates
Corning® Trypsin

2 Options Available Below

Product Details & Documents

0,05% Trypsin/0,53 mM EDTA in HBSS (Hank’s Balanced Salt Solution) with sodium bicarbonate, without calcium and magnesium.

  • Trypsin EDTA 1X 0,05% Trypsin/0,53 mM EDTA in HBSS with sodium bicarbonate, without calcium and magnesium
  • Porcine Parvovirus tested

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin. This product contains trypsin at a concentration of 0,05% with 0,53 mM EDTA, and does not contain sodium bicarbonate.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 25-052-CI
  • MDTC25-052-CV
  • Enzyme name:
  • Trypsin
  • Trypsin
  • CAS No.:
  • 9002-07-7
  • 9002-07-7

Specifications

Product Family Options

Product Information

  • Pk Info
    Pk
    Availability
    Price
  • 100 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - SFr. 81.60
    SFr. 81.60
    1 * 6 ST
       SFr. 81.60

    Specifications:
    More
    Specifications
    Cat. No.
    25-052-CI
    Enzyme name
    Trypsin
    CAS No.
    9002-07-7
  • 500 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - SFr. 293.00
    SFr. 293.00
    1 * 6 ST
       SFr. 293.00

    Specifications:
    More
    Specifications
    Cat. No.
    MDTC25-052-CV
    Enzyme name
    Trypsin
    CAS No.
    9002-07-7
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..