Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(+)-Glucose solution 45%, Corning®

D-(+)-Glucose solution 45%, Corning®

Plastic bottle, 1×100 ml, 100 mL
Catalog # 25-037-CI
Supplier:   Corning
D-(+)-Glucose solution 45%, Corning®
Catalog # 25-037-CI
Supplier:  Corning
CAS Number:  50-99-7
D-(+)-Glucose solution 45%, Corning®
D-(+)-Glucose solution 45%, Corning®

Plastic bottle, 1×100 ml, 100 mL

CAS Number:  50-99-7
Price
SFr. 44.40
Log in for availability
1 * 100 mL
Product Details & Documents

45% Glucose Solution. Glucose is a main source of energy for living organisms and is used as a nutrient source in cell culture media. This 45% (w/v) solution has been to ensure no cytotoxicity.

Synonyms:  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  • CAS Number:50-99-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148912
  • Molecular Weight:180.16 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • pH:
    3.5 ± 0.5 @ 20-25°
  • Osmolality:
    250-280 mOsm/Kg H20 @ 1:10
  • Endotoxin:
    ≤ 0.25 EU/ml @ FW
  • Sterility :
    Pass
  • Cytotoxicity:
    Pass

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Pk Info:
    1×100 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.