Cleanroom mops, MicroMopp Plus
PPSA2700530
- Description:MicroMop Plus, pre-washed, 130 g
- Length:400 mm
- Pk:1
Mops for wet and moist cleaning in single-stage technique.
- For all smooth and hard floor coverings
- Made up of high-tech-microfiber 100% PES
- With backfilling, cover sheet, loop with TEC-polyester and pocket of harness for extremely tear-proof
For the use in ISO class 9.