ContactUsLinkComponent
Fat test strips, (LRSM)
Fat test strips, (LRSM)
Catalog # MMMAFETT200
Supplier:  3M
CAS Number:  
undefined
Fat test strips, (LRSM)
Catalog # MMMAFETT200
Supplier:  3M
CAS Number:  
Specifications
  • Pk:
    200
  • Description:
    Fat test strips, 10 tubes/pack
  • Operating temperature:
    up to 180 °C
  • Test time:
    15 - 30 second
  • Test method:
    Free fatty acid
  • Tests:
    20
Specifications
About this item
The fat test strip is to determine fat quality by measuring the free fatty acids.
  • Operation at temperatures up to 180 °C
  • Easy and fast solution method
This rapid test is suitable for use with the same fat and the same fried food. The result can be easily read in approx. 15 to 30 seconds, by means of a colour change.