Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Calcium Standards
Calcium standard for AAS
Calcium standard for AAS
Calcium standard for AAS
ACSDA109.5NP.L1
:
CPACHEM
:
Calcium standard for AAS
ACSDA109.5NP.L1
:
CPACHEM
:
Concentration:
10000 mg/L
Matrix:
2-5% HNO₃
Pk:
100 mL
Environmentally Preferable:
Element:
Calcium
Symbol:
Ca