Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydroxide 1 mol/l (1 N) volumetric solution
Sodium hydroxide 1 mol/l (1 N) volumetric solution
Supplier:  PanReac AppliChem
Certificates
undefined
Sodium hydroxide 1 mol/l (1 N) volumetric solution
Supplier:  PanReac AppliChem

Specification Test Results

  • Appearance:
    Clear, colourless liquid
  • Titer at 20°C:
    0,999 - 1,001
  • Traceability:
    NIST
  • Solubility:
    Miscible with water
  • pH:
    13,7

Specification Test Results

Frequently Bought Together