ContactUsLinkComponent
Water samplers, SP 5 series
Water samplers, SP 5 series
Supplier:  MAXX
Certificates
undefined
Water samplers, SP 5 series
Supplier:  MAXX
Specifications
  • Catalog No:
  • 701-0144
  • 701-0151
  • 701-0104
  • 701-0102
  • 701-0154
  • 701-0139
  • 701-0150
  • 701-0153
  • 701-0098
  • 701-0147
  • 701-0101
  • 701-0148
  • 701-0149
  • 701-0152
  • 701-0141
  • 701-0075
  • 701-0142
  • 701-0146
  • 701-0100
  • 701-0143
  • 701-0145
  • 701-0155
  • 701-0105
  • 701-0140
  • 701-0103
  • 701-0099
  • Model:
  • SP5 S
  • SP5 B
  • SP5 B
  • SP5 S
  • SP5 B
  • SP5 B
  • SP5 S
  • SP5 S
  • SP5 S
  • SP5 B
  • SP5 S
  • SP5 S
  • SP5 S
  • SP5 S
  • SP5 S
  • SP5 B
  • SP5 S
  • Sample size:
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • 20 – 350 ml
  • Programs:
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • Datalog:
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
  • Accuracy:
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • Signal input:
  • 2x analogue; 8x digital
  • 2x analogue; 8x digital
  • 2x analogue; 8x digital
  • 2x analogue; 8x digital
  • 2x analogue; 8x digital
  • 2x analogue; 8x digital
  • Signal output:
  • Up to 8x digital
  • Up to 8x digital
  • Up to 8x digital
  • Up to 8x digital
  • Up to 8x digital
  • Communication interface:
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
  • Interfaces:
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
  • Control:
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
  • Suction height:
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • Power requirements:
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • 8 – 16 V
  • W×D×H:
  • 690×645×1290 mm
  • 760×745×1100 mm
  • 760×745×1100 mm
  • 690×645×1290 mm
  • 760×745×1100 mm
  • 760×745×1100 mm
  • 690×645×1290 mm
  • 690×645×1290 mm
  • 690×645×1290 mm
  • 760×745×1100 mm
  • 690×645×1290 mm
  • 690×645×1290 mm
  • 690×645×1290 mm
  • 690×645×1290 mm
  • 690×645×1290 mm
  • 760×745×1100 mm
  • 690×645×1290 mm
  • Weight:
  • 100 kg
  • 75 kg
  • 75 kg
  • 100 kg
  • 75 kg
  • 75 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 75 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 75 kg
  • 100 kg
Specifications
Sample container
Product Information