Banded earplugs, E-A-Rcap™
Banded earplugs, Blue/yellow
Catalog # 111-0912
Supplier: 3M
Jump to:
Product Details & Documents
Banded earplugs are a practical option and easy to use.
- Extremely lightweight, virtually no feeling of pressure
- Plugs do not push into your ear canal; prevents irritation
- Ideal for frequent insertion and removal
They can be worn around the neck when not needed, making them ideal for temporary use.
SNR: 23 dB
Certifications: EN 352-2
Specifications
- Colour:Blue/yellow
- Description:Banded earplugs
Accessories
Product Information
- Catalog #ColourAvailabilityPrice
- Replacement plugs for banded earplugs E-A-Rcap™/E-A-R™ Flexicap™Replacement plugs for banded earplugs E-A-Rcap™/E-A-R™ Flexicap™111-0446YellowLog in for availability1 * 50 PAIRE - SFr. 139.00
Product Information
- Catalog #ColourAvailabilityPrice
- Replacement plugs for banded earplugs E-A-Rcap™/E-A-R™ Flexicap™Replacement plugs for banded earplugs E-A-Rcap™/E-A-R™ Flexicap™111-0445YellowLog in for availability1 * 10 PAIRE - SFr. 36.80