Order Entry
Switzerland
ContactUsLinkComponent
Rubbermaid® Mobile Barrier
Rubbermaid® Mobile Barrier
Catalog # 116-0070
undefined
Rubbermaid® Mobile Barrier
Catalog # 116-0070

Specifications

  • Description:
    Portable mobile barrier, yellow, up to 4 m
  • Dimensions:
    600×350×1000 mm
  • Material:
    Polyethylene
  • Pk:
    1

Specifications

About this item

The mobile barrier is easy to use, easy to see, easy to move, and easy to store.

  • Flexible panels allow straight, curved, and circular configurations
  • Two non-marking wheels and four locking straps provide mobility and stability
  • Lightweight and compact (−15 kg)
  • Shipped assembled

Designed for indoor use, the mobile barrier extends to 4 metres with 16 articulating panels.