ESD occupational shoes, slip-on, Air Cushion, 32370
Supplier: Abeba
Certificates
About this item
Extremely light ESD shoe made from microfibre with a PU sole resistant to hydrocarbons.
- Air-cushioned PU walking sole with high slip resistance
- Instep area with elastic band
- Sanitized® treatment reduces odour and bacterial growth
Colour: White
Colour: White
Certifications: EN 61340 (ESD); EN ISO 20347:2012; O1, SRA
Frequently Bought Together