Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-AMPK gamma 1/2/3 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-9446R-CY3
Supplier:  Bioss
Anti-AMPK gamma 1/2/3 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-9446R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5' AMP-activated protein kinase gamma 1/2/3
  • Antigen symbol:
    AMPK gamma 1/2/3
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5571, 51422, 53632,
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    280-330/569
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

AMPK is a heterotrimeric complex comprising a catalytic a subunit and regulatory b and g subunits. It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways. AMPK is activated by high AMP and low ATP through a mechanism involving allosteric regulation, promotion of phosphorylation by an upstream protein kinase known as AMPK kinase and inhibition of dephosphorylation. Activated AMPK can phosphorylate and regulate in vivo hydroxy-methylglutaryl-CoA reductase and acetyl-CoA carboxylase, which are key regulatory enzymes of sterol synthesis and fatty acid synthesis, respectively. The human AMPKa1 and AMPKa2 genes encode 548 amino acid and 552 amino acid proteins, respectively. Human AMPKb1 encodes a 271 amino acid protein and human AMPKb2 encodes a 272 amino acid protein. The human AMPKg1 gene encodes a 331 amino acid protein. Human AMPKg2 and AMPKg3, which are 569 and 492 amino acid proteins, respectively, contain unique N-terminal domains and may participate directly in the binding of AMP within the AMPK complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMPK gamma 1/2/3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat