Order Entry
Switzerland
ContactUsLinkComponent
Anti-GNAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-GNAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-8160R-A488
Supplier:  Bioss
Anti-GNAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-8160R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Guanine Nucleotide Binding Protein (G Protein) Alpha Transducing Activity Polypeptide 1
  • Antigen symbol:
    GNAT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2779
  • Antigen synonyms:
    CSNB1G|CSNBAD3|GBT1|GNATR
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

GTP-binding proteins (G-proteins)are a family of heterotrimeric proteins that play a critical role in signal transduction by coupling cell surface, 7-transmembrane domain receptors to intracellular signaling pathways including second messenger generation (such as cyclic AMP, calcium and diacylglycerol), protein phosphorylation, ion channel activation, gene induction, cell growth and differentiation. Receptor activation catalyzes the exchange of GTP for GDP bound to the inactive G protein alpha subunit resulting in a conformational change and dissociation of the complex. The G protein alpha and beta-gamma subunits are capable of regulating various cellular effectors. Activation is terminated by a GTPase intrinsic to the G-alpha subunit.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G protein alpha
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat