Order Entry
Switzerland
ContactUsLinkComponent
Anti-CMTM8 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-8024R-CY5.5
Anbieter:  Bioss
Anti-CMTM8 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-8024R-CY5.5
Anbieter:  Bioss

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    CKLF-like MARVEL transmembrane Domain Containing 8
  • Antigen symbol:
    CMTM8
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    152189
  • Antigen synonyms:
    CKLFSF8-V2|CKLFSF8
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Technische Daten

Über diesen Artikel

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CMTM8
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: