Anti-HAX1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-7626R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HCLS1 associated protein X-1
- Antigen symbol:HAX1
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10456
- Antigen synonyms:SCN3|HS1BP1|HCLSBP1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
RelevanceHAX1 associates with HS1, binding to its N-terminal region. It is also known to associate with PKD2 (involved in polycystic kidney disease) and with cortactin/EMS1. HAX1 is also reported to bind to hairpin structures in vimentin and DNA polymerase beta mRNAs, so may play a role in mRNA stability and transport. It may also function in promoting cell survival. Defects in HAX1 are the cause of autosomal recessive severe congenital neutropenia 3 (SCN3) also called Kostmann disease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HAX1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat