Anti-HAX1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-7626R-A555
: Bioss
- Antikörper-Typ:Primär
- Antigen-Name:HCLS1 associated protein X-1
- Antigen-Symbol:HAX1
- Klonalität:Polyklonal
- Konjugation:Alexa Fluor® 555
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Kreuzadsorption:No
- Form:liquid
- Gen-ID:10456
- Antigen-Synonyme:SCN3|HS1BP1|HCLSBP1
- Lagerungspuffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Lagertemperatur:Store at –20 °C for 12 months
- Konzentration:1 μg/μl
- Transporttemperatur:4°C
- Reinigung:Purified by Protein A
- VE:100 µl
RelevanceHAX1 associates with HS1, binding to its N-terminal region. It is also known to associate with PKD2 (involved in polycystic kidney disease) and with cortactin/EMS1. HAX1 is also reported to bind to hairpin structures in vimentin and DNA polymerase beta mRNAs, so may play a role in mRNA stability and transport. It may also function in promoting cell survival. Defects in HAX1 are the cause of autosomal recessive severe congenital neutropenia 3 (SCN3) also called Kostmann disease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HAX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat