- Antibody type:Primary
- Antigen name:Nucleolar Protein 3
- Antigen symbol:NOL3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8996
- Antigen synonyms:FCM|NOP|Apoptosis repressor with CARD|Myp|MYP|NOP30|Nucleolar protein 3|ARC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD)bcontaining molecules and a caspase family of proteases. CARD containing cell death regulators include RAIDD, RICK, BCL10, Apaf 1, caspase 9 and caspase 2. Apoptosis repressor with CARD is a CARD domain containing protein that interacts with caspase 2 and 8 to inhibit enzymatic activity of caspase 8. Apoptosis repressor with CARD suppresses apoptosis induced by cell death adapters FADD and TRADD and by cell death receptors Fas, TNFR 1, and DR3. The mRNA of Apoptosis repressor with CARD is primarily expressed in skeletal muscle and cardiac tissue. The nuclear isoform (1/Nop30) may be involved in RNA splicing and the cytoplasmic isoform (2/Myp) may inhibit apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NOL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat