Order Entry
Switzerland
ContactUsLinkComponent
Anti-CA8 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-6516R-CY3
 :  Bioss
Anti-CA8 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-6516R-CY3
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Carbonic Anhydrase 8
  • Antigen symbol:
    CA8
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    767
  • Antigen synonyms:
    CALS|CA-VIII|CARP|CAMRQ3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CA8
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat