Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-Quiescin Q6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalognr. BOSSBS-6394R-FITC
Anbieter:  Bioss
Anti-Quiescin Q6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalognr. BOSSBS-6394R-FITC
Anbieter:  Bioss

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Quiescin Q6
  • Klonalität:
    Polyklonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    5768
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Technische Daten

Über diesen Artikel

This gene encodes a protein that contains domains of thioredoxin and ERV1, members of two long-standing gene families. The gene expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. Two transcript variants encoding two different isoforms have been found for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Quiescin Q6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat