Order Entry
Switzerland
ContactUsLinkComponent
Anti-SYK Rabbit Polyclonal Antibody (Cy3®)
Anti-SYK Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6127R-CY3
Supplier:  Bioss
Anti-SYK Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6127R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Spleen Tyrosine Kinase
  • Antigen symbol:
    SYK
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    6850
  • Antigen synonyms:
    Tyrosine-protein kinase SYK|Spleen tyrosine kinase|SYK|p72-Syk
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Catalyzes the specific attachment of an amino acid to its cognate tRNA in a 2 step reaction: the amino acid (AA) is first activated by ATP to form AA-AMP and then transferred to the acceptor end of the tRNA. When secreted, acts as a signaling molecule that induces immune response through the activation of monocyte/macrophages. Catalyzes the synthesis of diadenosine oligophosphate (Ap4A), a signaling molecule involved in the activation of MITF transcriptional activity. Interacts with HIV-1 virus GAG protein, facilitating the selective packaging of tRNA(3)(Lys), the primer for reverse transcription initiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYK
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat