Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-MFSD2A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-6073R-FITC
Supplier:  Bioss
Anti-MFSD2A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-6073R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Major Facilitator Superfamily Domain Containing 2a
  • Antigen symbol:
    MFSD2A
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    84879
  • Antigen synonyms:
    ZGC,101615|Major facilitator superfamily domain-containing protein 2A-A|mfsd2a
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    345-395/543
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Sodium-dependent lysophosphatidylcholine (LPC) symporter, which plays an essential role for blood-brain barrier formation and function. Specifically expressed in endothelium of the blood-brain barrier of micro-vessels and transports LPC into the brain. Transport of LPC is essential because it constitutes the major mechanism by which docosahexaenoic acid (DHA), an omega-3 fatty acid that is essential for normal brain growth and cognitive function, enters the brain. Transports LPC carrying long-chain fatty acids such LPC oleate and LPC palmitate with a minimum acyl chain length of 14 carbons. Does not transport docosahexaenoic acid in unesterified fatty acid. Specifically required for blood-brain barrier formation and function, probably by mediating lipid transport. Not required for central nervous system vascular morphogenesis (By similarity). Acts as a transporter for tunicamycin, an inhibitor of asparagine-linked glycosylation. In placenta, acts as a receptor for ERVFRD-1/syncytin-2 and is required for trophoblast fusion (PubMed:18988732).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MFSD2A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat