Anti-PBOV1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-5939R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Prostate And breast cancer overexpressed 1
- Antigen symbol:PBOV1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:59351
- Antigen synonyms:PBOV-1|Prostate and breast cancer overexpressed 1|UROC28|UC28.PBOV1_HUMAN|Prostate and breast cancer overexpressed gene 1 protein|PBOV 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
This gene is overexpressed in prostate and breast cancer.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PBOV1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human