Anti-AZU1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-5736R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Azurocidin 1
- Antigen symbol:AZU1
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:566
- Antigen synonyms:AZAMP|HBP|AZU|NAZC|HUMAZUR|CAP37
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
TACC2 is a conserved family of centrosome and microtubule interacting proteins that are implicated in cancer. This gene encodes a protein that concentrates at centrosomes throughout the cell cycle. This gene lies within a chromosomal region associated with tumorigenesis. Expression of this gene is thought to affect the progression of breast tumors. Expression of this gene is also induced by erythropoietin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AZU1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat