Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-NL1 Rabbit Polyclonal Antibody (Cy3®)
Anti-NL1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-4212R-CY3
Supplier:  Bioss
Anti-NL1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-4212R-CY3
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Neuroligin 1
  • Antigen-Symbol:
    NL1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Cy3®
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    22871
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Immunogen:
    716-766/863
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Specifications

About this item

Cell surface protein involved in cell-cell-interactions via its interactions with neurexin family members. Plays a role in synapse function and synaptic signal transmission, and probably mediates its effects by recruiting and clustering other synaptic proteins. May promote the initial formation of synapses, but is not essential for this. In vitro, triggers the de novo formation of presynaptic structures. May be involved in specification of excitatory synapses (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neuroligin 1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat