Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-ADCY7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BOSSBS-3924R-HRP
 :  Bioss
Anti-ADCY7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BOSSBS-3924R-HRP
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Adenylate Cyclase 7
  • Antigen symbol:
    ADCY7
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    113
  • Antigen synonyms:
    AC7
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    905-955/1080
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

This gene encodes a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterized by the presence of twelve membrane-spanning domains in its sequences. [provided by RefSeq, Jul 2008].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ADCY7
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat