Order Entry
Switzerland
ContactUsLinkComponent
Anti-HINT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-HINT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-10228R-A350
Supplier:  Bioss
Anti-HINT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-10228R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Histidine triad nucleotide binding Protein 1
  • Antigen symbol:
    HINT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    HINT1|PKCI1|Protein kinase C inhibitor 1|NMAN|Histidine triad nucleotide-binding protein 1|PRKCNH1|PKCI-1|HINT
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The protein encoded by this gene can hydrolyze substrates such as AMP-morpholidate, AMP-N-alanine methyl ester, AMP-alpha-acetyl lysine methyl ester, and AMP-NH2. The encoded protein interacts with these substrates via a histidine triad motif, which is part of the loop that binds to the substrate. This gene has been found to be a tumor suppressing gene. Several transcript variants, but only one of them protein-coding, have been found for this gene. [provided by RefSeq, Dec 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HINT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat