Order Entry
Switzerland
ContactUsLinkComponent
Anti-DRD1 Rabbit Polyclonal Antibody (Cy3®)
Anti-DRD1 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-1007R-CY3
 :  Bioss
Anti-DRD1 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-1007R-CY3
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    dopamine receptor D1
  • Antigen symbol:
    DRD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1812
  • Antigen synonyms:
    DRD1A|DADR
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    155-192/446
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Dopamine receptor whose activity is mediated by G proteins which activate adenylyl cyclase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DRD1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat