Order Entry
Switzerland
ContactUsLinkComponent
Anti-FHIT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Katalognr. BOSSBS-3146R-A555
Anbieter:  Bioss
Anti-FHIT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Katalognr. BOSSBS-3146R-A555
Anbieter:  Bioss

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    Fragile Histidine Triad
  • Antigen symbol:
    FHIT
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2272
  • Antigen synonyms:
    AP3Aase|FHIT |AP3AaseAP3A hydrolase|FRA3B
  • Modification:
    phospho-Tyr114
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    90-130/147
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Technische Daten

Über diesen Artikel

Cleaves P(1)-P(3)-bis(5'-adenosyl) triphosphate (Ap3A) to yield AMP and ADP. Can also hydrolyze P(1)-P(4)-bis(5'-adenosyl) tetraphosphate (Ap4A), but has extremely low activity with ATP. Modulates transcriptional activation by CTNNB1 and thereby contributes to regulate the expression of genes essential for cell proliferation and survival, such as CCND1 and BIRC5. Plays a role in the induction of apoptosis via SRC and AKT1 signaling pathways. Inhibits MDM2-mediated proteasomal degradation of p53/TP53 and thereby plays a role in p53/TP53-mediated apoptosis. Induction of apoptosis depends on the ability of FHIT to bind P(1)-P(3)-bis(5'-adenosyl) triphosphate or related compounds, but does not require its catalytic activity, it may in part come from the mitochondrial form, which sensitizes the low-affinity Ca(2+) transporters, enhancing mitochondrial calcium uptake. Functions as tumor suppressor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FHIT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human