- Antibody type:Primary
- Antigen name:5' AMP-activated protein kinase beta 1 And 2
- Antigen symbol:AMPK beta 1 + 2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5565, 5564
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:220-270/272
- Purification:Purified by Protein A
- Pk:100 µl
PRKAB2 ans PRKAB1 are regulatory subunits of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status and plays a role in protecting cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways. AMPK is responsible for the regulation of fatty acid synthesis by phosphorylation of acetyl-CoA carboxylase (ACC). It also regulates cholesterol synthesis via phosphorylation and inactivation of hydroxymethylglutaryl-CoA reductase (HMGCR) and hormone-sensitive lipase. PRKAB2 may be a positive regulator of AMPK activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AMPK beta 1 + 2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat