Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-GRM2 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-GRM2 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-2139R-CY5.5
Supplier:  Bioss
Anti-GRM2 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-2139R-CY5.5
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Glutamate Receptor, Metabotropic 2
  • Antigen-Symbol:
    GRM2
  • Klonalität:
    Polyklonal
  • Konjugation:
    Cy5.5®
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    2912
  • Antigen-Synonyme:
    GPRC1B|Metabotropic glutamate receptor 2|GRM2|mGlu2|MGLUR2|GLUR2
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Specifications

About this item

L glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GRM2+GRM4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat