Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-PDEF/SPDEF Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-PDEF/SPDEF Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1866R-A647
Supplier:  Bioss
Anti-PDEF/SPDEF Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1866R-A647
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    PDEF/SPDEF
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 647
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    25803
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Specifications

About this item

SPDEF, also known as PDEF is an ETS transcription factor expressed in the prostate epithelial cells. It is thought to act as an androgen-independent transactivator of the prostate-specific antigen (PSA) promoter and also as a transcriptional activator for the SERPINB5 promoter. It is highly expressed in prostate carcinoma cells and is thought to be involved in the regulation of the prostate gland and/or prostate cancer development. It is also expressed in epithelial breast carcinoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDEF/SPDEF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat